missing translation for 'onlineSavingsMsg'
Learn More

3-Hepten-2-one, 95%

Product Code. 11459386
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459386 50 g 50g
11449386 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11459386 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A19086.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 1119-44-4 | C7H12O | 112.17 g/mol

It is used in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews). It is also used to produce Heptan-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews). It is also used to produce Heptan-2-one.

Solubility
Soluble in alcohol and slightly soluble in water(3087 mg/L @ 25°C (est.))

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1119-44-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00015564
InChI Key JHHZQADGLDKIPM-AATRIKPKSA-N
Synonym 3-hepten-2-one, butylideneacetone, e-hept-3-en-2-one, 3e-3-hepten-2-one, methyl pentenyl ketone, e-3-hepten-2-one, 3-hepten-2-one, e, unii-tk02t5fjdo, tk02t5fjdo, 3e-hept-3-en-2-one
PubChem CID 5364578
SMILES CCC\C=C\C(C)=O

Specifikationer

Density 0.86
Boiling Point 156°C
Flash Point 49°C (120°F)
Refractive Index 1.445
Quantity 50 g
UN Number UN1224
Beilstein 1739239
Solubility Information Soluble in alcohol and slightly soluble in water(3087mg/L @ 25°C (est.))
IUPAC Name (3E)-hept-3-en-2-one
Formula Weight 112.16
Percent Purity 97%
Chemical Name or Material 3-Hepten-2-one
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.