missing translation for 'onlineSavingsMsg'
Learn More

3-Heptanol, 98%

Product Code. 15304978 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15304978 25 g 25g
15314978 100 g 100g
15324978 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15304978 Supplier Thermo Scientific Acros Supplier No. 149090250
  • ONLINE EXCLUSIVE PRICE
    £33.10
    £29.65 / 25g
    Save £3.45
    10% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 589-82-2 | C7H16O | 116.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-82-2
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.2
InChI Key RZKSECIXORKHQS-UHFFFAOYSA-N
Synonym 3-heptanol, 3-hydroxyheptane, ethyl butyl carbinol, butyl ethyl carbinol, butylethylcarbinol, xi-3-heptanol, fema no. 3547, ethylbutylcarbinol, heptanol-3, 3-heptyl alcohol
PubChem CID 11520
IUPAC Name heptan-3-ol
SMILES CCCCC(CC)O

Specifications

Density 0.8180g/mL
Boiling Point 156.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3(CH2)3CH(OH)C2H5
Refractive Index 1.4214
Quantity 25 g
Beilstein 01,I,205
Specific Gravity 0.818
Solubility Information Solubility in water: insoluble.
Formula Weight 116.2
Percent Purity 98%
Chemical Name or Material 3-Heptanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Heptanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: 3-Heptanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.