missing translation for 'onlineSavingsMsg'
Learn More

3-Guanidinopropionic Acid, TRC

Product Code. 30390728
Change view
Click to view available options
Quantity:
2 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390728 5 g
30408653 2 g
30404471 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390728 Supplier Toronto Research Chemicals Supplier No. TRCG8357755G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-09-3
Molecular Formula C4 H9 N3 O2
Molecular Weight (g/mol) 131.13
Synonym 3-Guanidinopropanoic Acid, 3-Guanidinopropionic Acid, Guanidinepropanoic Acid, Guanidinepropionic Acid, Guanidinopropionic Acid, N-(3-Propanoic Acid)guanidine, PNU 10483, Propanoic Acid, 3-[(aminoiminomethyl)amino]-, β-Guanidinopropionic Acid, β-Guanidopropionic Acid
IUPAC Name 3-carbamimidamidopropanoic acid
SMILES NC(=N)NCCC(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H9N3O2/c5-4(6)7-2-1-3(8)9/h1-2H2,(H,8,9)(H4,5,6,7)
Formula Weight 131.0695
Chemical Name or Material Guanidinopropionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.