missing translation for 'onlineSavingsMsg'
Learn More

3-Furoic acid, 99%

Product Code. 11312537
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312537 5 g 5g
11322537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312537 Supplier Thermo Scientific Alfa Aesar Supplier No. L11629.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 488-93-7 | C5H4O3 | 112.08 g/mol

3-Furoic acid is used in synthesis of furan-2,5- and furan-2,4-dicarboxylic acid under solvent-free conditions via disproportionation reaction. It exhibits hypolipidemic activity in rodents. It lowers serum cholesterol and serum triglyceride levels in mice and rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Furoic acid is used in synthesis of furan-2,5- and furan-2,4-dicarboxylic acid under solvent-free conditions via disproportionation reaction. It exhibits hypolipidemic activity in rodents. It lowers serum cholesterol and serum triglyceride levels in mice and rats.

Solubility
Soluble in water, ether, alcohol, ethanol, and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-93-7
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
MDL Number MFCD00005350
InChI Key IHCCAYCGZOLTEU-UHFFFAOYSA-N
Synonym 3-furoic acid, 3-furancarboxylic acid, 3-carboxyfuran, 3-furanoic acid, unii-88a3s7rg98, 3-furoate, 3-furans, 3-furanoate, pubchem6945, acmc-209kee
PubChem CID 10268
ChEBI CHEBI:30846
IUPAC Name furan-3-carboxylic acid
SMILES OC(=O)C1=COC=C1

Specifications

Melting Point 120°C to 123°C
Quantity 5 g
Beilstein 108638
Solubility Information Soluble in water,ether,alcohol,ethanol,and methanol.
Formula Weight 112.08
Percent Purity 99%
Chemical Name or Material 3-Furoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.