missing translation for 'onlineSavingsMsg'
Learn More

3-Furanmethanol, TRC

Product Code. 30401641
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401641 50 mg
30405510 100 mg
30406454 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401641 Supplier Toronto Research Chemicals Supplier No. TRCF86400350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4412-91-3
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
Synonym 3-(Hydroxymethyl)furan, 3-Furancarbinol, 3-Furfuryl Alcohol, 3-Furylcarbinol, 3-Furylmethanol, 3-Furylmethyl Alcohol, Furan-3-ylmethanol
SMILES OCc1cocc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H6O2/c6-3-5-1-2-7-4-5/h1-2,4,6H,3H2
Formula Weight 98.0368
Chemical Name or Material 3-Furanmethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.