missing translation for 'onlineSavingsMsg'
Learn More

3-Furanmethanol, 99%

Product Code. 10085813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10085813 25 g Glass bottle 25g
10568873 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10085813 Supplier Thermo Scientific Acros Supplier No. 188080250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4412-91-3 | C5H6O2 | 98.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4412-91-3
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00005352
InChI Key STJIISDMSMJQQK-UHFFFAOYSA-N
Synonym 3-furanmethanol, furan-3-methanol, 3-furylmethanol, 3-hydroxymethyl furan, 3-furfuryl alcohol, 3-hydroxymethylfuran, 3-furyl alcohol, furan-3-yl-methanol, furan-3-yl methanol, 3-hydroxymethyl-furan
PubChem CID 20449
IUPAC Name furan-3-ylmethanol
SMILES OCC1=COC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1390g/mL
Boiling Point 79.0°C to 80.0°C (17.0 mmHg)
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4835 to 1.4855
Quantity 25 g
Beilstein 17, III, 1266
Specific Gravity 1.139
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 98.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Furanmethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Furanmethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.