missing translation for 'onlineSavingsMsg'
Learn More

3-Furancarboximidamide hydrochloride, Technical Grade, Thermo Scientific™

Product Code. 10120792
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10120792 1 g 1g
10110792 250 mg 250mg
10623645 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10120792 Supplier Thermo Scientific Maybridge Supplier No. MO00101DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 54649-21-7
Molecular Formula C5H7ClN2O
Molecular Weight (g/mol) 146.574
InChI Key RJLCJJIXGGSRPZ-UHFFFAOYSA-N
Synonym furan-3-carboximidamide hydrochloride, 3-furancarboximidamide hydrochloride, furan-3-carboxamidine hcl, furan-3-carboxamidine, chloride, 3-furancarboximidamide,hydrochloride 1:1
PubChem CID 2779666
IUPAC Name furan-3-carboximidamide;hydrochloride
SMILES C1=COC=C1C(=N)N.Cl

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 146.58
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Furancarboximidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.