missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Furancarboximidamide hydrochloride, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Quantity | 250 mg |
Formula Weight | 146.58 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 3-Furancarboximidamide hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'