missing translation for 'onlineSavingsMsg'
Learn More

3-Furancarbothioamide, 97%, Thermo Scientific™

Product Code. 10282692
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10282692 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10282692 Supplier Thermo Scientific Maybridge Supplier No. MO00105DA
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 59918-68-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H5NOS
MDL Number MFCD02179222
Quantity 1 g
Synonym 3-furancarbothioamide, 3-furancarbothioamide 9ci, amino-3-furylmethane-1-thione
InChI Key FHPWEMZQNZDEMS-UHFFFAOYSA-N
SMILES C1=COC=C1C(=S)N
IUPAC Name furan-3-carbothioamide
Molecular Weight (g/mol) 127.161
PubChem CID 2779671
Formula Weight 127.17
Percent Purity 97%
Chemical Name or Material 3-Furancarbothioamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.