missing translation for 'onlineSavingsMsg'
Learn More

3-Furaldehyde, 98%, stabilized

Product Code. 11327631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11327631 1 g Glass bottle 1g
11956171 5 g Glass bottle 5g
11966171 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327631 Supplier Thermo Scientific Acros Supplier No. 393260010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 498-60-2 | C5H4O2 | 96.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-60-2
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.08
InChI Key AZVSIHIBYRHSLB-UHFFFAOYSA-N
Synonym 3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948
PubChem CID 10351
ChEBI CHEBI:87609
IUPAC Name furan-3-carbaldehyde
SMILES C1=COC=C1C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.1500g/mL
Boiling Point 145.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.15
Formula Weight 96.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Furaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Furaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.