missing translation for 'onlineSavingsMsg'
Learn More

3-Formylsalicylic acid hydrate, 97%

Product Code. 11915131 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11915131

Brand: Thermo Scientific Acros 377070250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1266615-78-4 | C8H6O4 | 166.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-78-4
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD03804454
InChI Key YOEUNKPREOJHBW-UHFFFAOYSA-N
Synonym 3-Formyl-2-hydroxybenzoic acid hydrate, 2-Hydroxyisophthalaldehyde acid hydrate
IUPAC Name 3-formyl-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC(C=O)=C1O

Specifications

Melting Point 249°C to 252°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 25 g
Formula Weight 166.13
Percent Purity 97%
Chemical Name or Material 3-Formylsalicylic acid hydrate
compliance-icons compliance-icons
Product Identifier
  • 3-Formylsalicylic acid hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.