missing translation for 'onlineSavingsMsg'
Learn More

3-Formylphenylboronic Acid, TRC

Product Code. 30390155
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390155 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390155 Supplier Toronto Research Chemicals Supplier No. TRCF7009701G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Formylphenylboronic Acid
CAS 87199-16-4
Molecular Formula C7H7BO3
Quantity 1 g
Synonym B-(3-Formylphenyl)boronic Acid, (3-Formylbenzene)boronic Acid, m-Formylphenylboronic Acid
InChI Formula InChI=1S/C7H7BO3/c9-5-6-2-1-3-7(4-6)8(10)11/h1-5,10-11H
SMILES OB(O)c1cccc(C=O)c1
Molecular Weight (g/mol) 149.94
Formula Weight 150.0488
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.