missing translation for 'onlineSavingsMsg'
Learn More

3-Formylbenzonitrile, 97%, Thermo Scientific™

Product Code. 10012174
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10012174 25 g 25g
10565564 1 g 1g
10478665 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10012174 Supplier Thermo Scientific Maybridge Supplier No. SEW02565EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 24964-64-5
Molecular Formula C8H5NO
Molecular Weight (g/mol) 131.134
MDL Number MFCD00003344
InChI Key HGZJJKZPPMFIBU-UHFFFAOYSA-N
Synonym 3-cyanobenzaldehyde, m-cyanobenzaldehyde, benzonitrile, 3-formyl, m-formylbenzonitrile, m-cyanobenazldehyde, 3-cyano benzaldehyde, 3-formyl-benzonitrile, 3-formylbenzenecarbonitrile, 3cyano-benzaldehyde, m-cyano benzaldehyde
PubChem CID 90670
IUPAC Name 3-formylbenzonitrile
SMILES C1=CC(=CC(=C1)C=O)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula NCC6H4CHO
Quantity 25 g
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material 3-Formylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.