missing translation for 'onlineSavingsMsg'
Learn More

3-Formylbenzeneboronic acid

Product Code. 11477116
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477116 25 g 25g
11457116 1 g 1g
11467116 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477116 Supplier Thermo Scientific Alfa Aesar Supplier No. B25437.14
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 87199-16-4 | C7H7BO3 | 149.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87199-16-4
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00161356
InChI Key HJBGZJMKTOMQRR-UHFFFAOYSA-N
Synonym 3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
PubChem CID 2734356
IUPAC Name (3-formylphenyl)boronic acid
SMILES OB(O)C1=CC=CC(C=O)=C1

Specifications

Melting Point 109°C to 113°C
Quantity 25 g
Beilstein 3030769
Sensitivity Air Sensitive
Formula Weight 149.94
Chemical Name or Material 3-Formylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.