missing translation for 'onlineSavingsMsg'
Learn More

3-(Formylamino)-1H-pyrazole-4-carboxamide, TRC

Product Code. 17992405
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992405 250 mg 250mg
17955035 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17992405 Supplier Toronto Research Chemicals Supplier No. TRCF696750250MG
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22407-20-1
Molecular Formula C5 H6 N4 O2

Specifications

Quantity 250 mg
Synonym 1H-Pyrazole-4-carboxamide, 3-(formylamino)-, Pyrazole-4-carboxamide, 3-formamido- (8CI)
InChI Formula InChI=1S/C5H6N4O2/c6-4(11)3-1-8-9-5(3)7-2-10/h1-2H,(H2,6,11)(H2,7,8,9,10)
SMILES NC(=O)c1cn[nH]c1NC=O
IUPAC Name 5-formamido-1H-pyrazole-4-carboxamide
Molecular Weight (g/mol) 154.13
Formula Weight 154.0491
Percent Purity >95%
Chemical Name or Material 5-(formylamino)-1H-pyrazole-4-carboxamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.