missing translation for 'onlineSavingsMsg'
Learn More

3-Formyl-1H-indole-7-carboxylic acid, ≥95%, Thermo Scientific™

Product Code. 10485132
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10485132 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10485132 Supplier Thermo Scientific Maybridge Supplier No. SB02107DA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 317854-65-2
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C10H7NO3
MDL Number MFCD00550642
Quantity 1 g
Synonym 7-carboxy-3-formyl-1h-indole, 3-formylindole-7-carboxylic acid, 1h-indole-7-carboxylicacid, 3-formyl, 1h-indole-7-carboxylic acid, 3-formyl, 3-formyl-indole-7-carboxylic acid, 7-carboxy-1h-indole-3-carboxaldehyde
InChI Key HABYJDPNVBUDGD-UHFFFAOYSA-N
SMILES OC(=O)C1=C2NC=C(C=O)C2=CC=C1
IUPAC Name 3-formyl-1H-indole-7-carboxylic acid
Molecular Weight (g/mol) 189.17
PubChem CID 2796697
Formula Weight 189.17
Percent Purity 95%
Chemical Name or Material 3-Formyl-1H-indole-7-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.