missing translation for 'onlineSavingsMsg'
Learn More

3'-Formyl(1,1'-biphenyl)-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10095452
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10095452 1 g 1g
10679724 250 mg 250mg
10117774 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10095452 Supplier Thermo Scientific Maybridge Supplier No. MO01257DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 222180-19-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD04039068
InChI Key QOCUSJPHQSKJJR-UHFFFAOYSA-N
Synonym 3'-formyl-1,1'-biphenyl-3-carboxylic acid, 3-3-formylphenyl benzoic acid, 3'-formylbiphenyl-3-carboxylic acid, 3'-formyl 1,1'-biphenyl-3-carboxylic acid, 3'-formyl-biphenyl-3-carboxylic acid, amtda124, acmc-1cb16, 1,1'-biphenyl-3-carboxylicacid, 3'-formyl
PubChem CID 2794721
IUPAC Name 3-(3-formylphenyl)benzoic acid
SMILES C1=CC(=CC(=C1)C=O)C2=CC(=CC=C2)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 3'-Formyl(1, 1'-biphenyl)-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.