missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorosalicylaldehyde, 98%

Product Code. 11344246
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344246 5 g 5g
11354246 250 mg 250mg
11334246 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344246 Supplier Thermo Scientific Alfa Aesar Supplier No. L19324.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 394-50-3 | C7H5FO2 | 140.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-50-3
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
MDL Number MFCD00003319
InChI Key NWDHTEIVMDYWQJ-UHFFFAOYSA-N
Synonym 3-fluorosalicylaldehyde, 3-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-2-hydroxy, zlchem 257, pubchem4215, 3-fluor-salicylaldehyd, acmc-209j5k, ksc497m8b, 2-hydroxy-3-fluoro-benzaldehyde
PubChem CID 587788
IUPAC Name 3-fluoro-2-hydroxybenzaldehyde
SMILES OC1=C(C=O)C=CC=C1F

Specifications

Melting Point 67°C to 71°C
Quantity 5 g
Beilstein 1932806
Sensitivity Air Sensitive
Formula Weight 140.12
Percent Purity 98%
Chemical Name or Material 3-Fluorosalicylaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.