missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine, 98%

Product Code. 11409203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409203 1 g 1g
11419203 5 g 5g
11429203 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409203 Supplier Thermo Scientific Alfa Aesar Supplier No. A11693.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 372-47-4 | C5H4FN | 97.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-47-4
Molecular Formula C5H4FN
Molecular Weight (g/mol) 97.092
MDL Number MFCD00006374
InChI Key CELKOWQJPVJKIL-UHFFFAOYSA-N
Synonym pyridine, 3-fluoro, 3-fluorpyridin, ccris 1717, 3-fluropyridine, 3-fluoro-pyridine, 3-fluoro-pyridinium, pubchem1095, acmc-1coty, 3-fluoropyridine, ksc173o1l
PubChem CID 67794
IUPAC Name 3-fluoropyridine
SMILES C1=CC(=CN=C1)F

Specifications

Boiling Point 106°C to 108°C
Flash Point 13°C (55°F)
Quantity 1 g
UN Number UN1993
Beilstein 105702
Formula Weight 97.09
Percent Purity 98%
Chemical Name or Material 3-Fluoropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.