missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine-2-carboxaldehyde, 98%

Product Code. 15488488
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488488 5 g 5g
15498488 250 mg 250mg
15478488 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15488488 Supplier Thermo Scientific Alfa Aesar Supplier No. H61902.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31224-43-8 | C6H4FNO | 125.102 g/mol

3-Fluoropyridine-2-carboxaldehyde is used to prepare taxoids derived from 9β-dihydrobaccatin-9,10-acetals with anti-tumor activities. It is also an intermediate used to synthesize S-3578 derivatives with potent activities against both methicillin-resistant Staphylococcus aureus (MRSA) and Pseudomonas aeruginosa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoropyridine-2-carboxaldehyde is used to prepare taxoids derived from 9β-dihydrobaccatin-9,10-acetals with anti-tumor activities. It is also an intermediate used to synthesize S-3578 derivatives with potent activities against both methicillin-resistant Staphylococcus aureus (MRSA) and Pseudomonas aeruginosa.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31224-43-8
Molecular Formula C6H4FNO
Molecular Weight (g/mol) 125.102
MDL Number MFCD07781234
InChI Key OZIMPUNGBUYCSP-UHFFFAOYSA-N
Synonym 3-fluoro-2-formylpyridine, 3-fluoropicolinaldehyde, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoropyridine-2-carboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 3-fluoro, 2-formyl-3-fluoropyridine, pubchem5142, acmc-209hkw, 3-fluoro-2-formyl-pyridine
PubChem CID 11344017
IUPAC Name 3-fluoropyridine-2-carbaldehyde
SMILES C1=CC(=C(N=C1)C=O)F

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 125.1
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Fluoropyridine-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.