Learn More
3-Fluoropyridine-2-carboxaldehyde, 97%, Maybridge™
Brand: Maybridge CC30904CB
Code : 31
Additional Details : CAS Number : 31224-43-8 Weight : 0.05025kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
31224-43-8 | |
125.102 | |
OZIMPUNGBUYCSP-UHFFFAOYSA-N | |
11344017 | |
C1=CC(=C(N=C1)C=O)F |
C6H4FNO | |
MFCD07781234 | |
3-fluoro-2-formylpyridine, 3-fluoropicolinaldehyde, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoropyridine-2-carboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 3-fluoro, 2-formyl-3-fluoropyridine, pubchem5142, acmc-209hkw, 3-fluoro-2-formyl-pyridine | |
3-fluoropyridine-2-carbaldehyde |
Specifications
3-Fluoropyridine-2-carboxaldehyde | |
250mg | |
96.5 | |
97% | |
MFCD07781234 | |
OZIMPUNGBUYCSP-UHFFFAOYSA-N | |
3-fluoropyridine-2-carbaldehyde | |
11344017 | |
97% |
Amber Glass Bottle | |
31224-43-8 | |
100.0 | |
C6H4FNO | |
3-fluoro-2-formylpyridine, 3-fluoropicolinaldehyde, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoropyridine-2-carboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 3-fluoro, 2-formyl-3-fluoropyridine, pubchem5142, acmc-209hkw, 3-fluoro-2-formyl-pyridine | |
C1=CC(=C(N=C1)C=O)F | |
125.102 | |
125.1 |