missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyrazin-2-amine, TRC

Product Code. 30398033
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398033 5 mg
30408461 10 mg
30405866 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398033 Supplier Toronto Research Chemicals Supplier No. TRCF4020285MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1206523-95-6
Molecular Formula C4H4N3F
Molecular Weight (g/mol) 113.09
SMILES Nc1nccnc1F

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C4H4FN3/c5-3-4(6)8-2-1-7-3/h1-2H,(H2,6,8)
Formula Weight 113.0389
Chemical Name or Material 3-Fluoropyrazin-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.