missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropropiophenone, TRC

Product Code. 30393642
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393642 500 mg
30390942 1 g
30400288 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393642 Supplier Toronto Research Chemicals Supplier No. TRCF595600500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-67-4
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
Synonym 1-(3-Fluorophenyl)-1-propanone, 3'-Fluoropropiophenone, 1-(3-Fluorophenyl)propan-1-one, Ethyl m-Fluorophenyl Ketone, m-Fluoropropiophenone
SMILES CCC(=O)c1cccc(F)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H9FO/c1-2-9(11)7-4-3-5-8(10)6-7/h3-6H,2H2,1H3
Formula Weight 152.0637
Chemical Name or Material 3-Fluoropropiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.