missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropropan-1-ol, TRC

Product Code. 30395628
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395628 10 g 10g
30405511 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395628 Supplier Toronto Research Chemicals Supplier No. TRCF59860510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Fluoropropan-1-ol
CAS 462-43-1
Molecular Formula C3H7FO
Quantity 10 g
Synonym 3-Fluoro-1-propanol, 3-Fluoropropanol, 3-Hydroxypropyl Fluoride
InChI Formula InChI=1S/C3H7FO/c4-2-1-3-5/h5H,1-3H2
SMILES OCCCF
IUPAC Name 3-fluoropropan-1-ol
Molecular Weight (g/mol) 78.09
Formula Weight 78.0481
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.