missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropiperidine hydrochloride, 97%

Product Code. 11309735
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11309735

Brand: Thermo Scientific Alfa Aesar L19776.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 737000-77-0 | C5H11ClFN | 139.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 737000-77-0
Molecular Formula C5H11ClFN
Molecular Weight (g/mol) 139.598
MDL Number MFCD03452799
InChI Key RDJUBLSLAULIAT-UHFFFAOYSA-N
Synonym 3-fluoropiperidine hydrochloride, 3-fluoropiperidine hcl, piperidine, 3-fluoro-, hydrochloride, pubchem24370, acmc-20an39, 3-fluoropiperidinehydrochloride, 3-fluoro-piperidine hydrochloride, 3-fluoranylpiperidine hydrochloride, 3-fluoro-piperidine, hydrochloride
PubChem CID 2759156
IUPAC Name 3-fluoropiperidine;hydrochloride
SMILES C1CC(CNC1)F.Cl

Specifications

Melting Point ∼227°C
Quantity 1 g
Beilstein 6145604
Sensitivity Hygroscopic
Formula Weight 139.6
Percent Purity 97%
Chemical Name or Material 3-Fluoropiperidine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.