missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropicolinonitrile, TRC

Product Code. 30401965
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401965 1 g
30406324 5 g
30404659 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401965 Supplier Toronto Research Chemicals Supplier No. TRCF6212201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97509-75-6
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.1
SMILES Fc1cccnc1C#N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H3FN2/c7-5-2-1-3-9-6(5)4-8/h1-3H
Formula Weight 122.028
Chemical Name or Material 3-Fluoropicolinonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.