missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophthalic acid, 97%, Thermo Scientific Chemicals

Catalog Number 11480244
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1583-67-1
C8H5FO4
184.122
MFCD00039524
BBCQSMSCEJBIRD-UHFFFAOYSA-N
3-fluorobenzene-1,2-dicarboxylic acid, 3-fluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylic acid, 3-fluoro, 1,2-benzenedicarboxylic acid, fluoro, 3-fluorophthalic, 3-fluorophtalic acid, 3-fluorophthatic acid, pubchem1405, 3-fluorophthalicacid
345381
3-fluorophthalic acid
C1=CC(=C(C(=C1)F)C(=O)O)C(=O)O
This item is not returnable. View return policy
1583-67-1
C8H5FO4
184.122
MFCD00039524
BBCQSMSCEJBIRD-UHFFFAOYSA-N
3-fluorobenzene-1,2-dicarboxylic acid, 3-fluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylic acid, 3-fluoro, 1,2-benzenedicarboxylic acid, fluoro, 3-fluorophthalic, 3-fluorophtalic acid, 3-fluorophthatic acid, pubchem1405, 3-fluorophthalicacid
345381
3-fluorophthalic acid
C1=CC(=C(C(=C1)F)C(=O)O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼170°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 2616684
Formula Weight 184.12
Percent Purity 97%
Chemical Name or Material 3-Fluorophthalic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'