Learn More
3-Fluorophenylboronic acid, 97%, Thermo Scientific Chemicals
£29.20 - £334.00
Chemical Identifiers
CAS | 768-35-4 |
---|---|
Molecular Formula | C6H6BFO2 |
Molecular Weight (g/mol) | 139.92 |
MDL Number | MFCD00236042 |
InChI Key | KNXQDJCZSVHEIW-UHFFFAOYSA-N |
Synonym | 3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol, 3-fluorophenylbornic acid, fpba, boronic acid, b-3-fluorophenyl, 3-fluoro-phenyl-boronic acid |
PubChem CID | 2733986 |
IUPAC Name | (3-fluorophenyl)boronic acid |
SMILES | OB(O)C1=CC=CC(F)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10526261
|
Thermo Scientific Acros
345640010 |
1 g | Glass bottle |
£29.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10683172
|
Thermo Scientific Acros
345640050 |
5 g | Glass bottle |
£94.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10297503
|
Thermo Scientific Acros
345640250 |
25 g | Glass bottle |
£334.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
768-35-4 | |
139.92 | |
KNXQDJCZSVHEIW-UHFFFAOYSA-N | |
2733986 | |
OB(O)C1=CC=CC(F)=C1 |
C6H6BFO2 | |
MFCD00236042 | |
3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol, 3-fluorophenylbornic acid, fpba, boronic acid, b-3-fluorophenyl, 3-fluoro-phenyl-boronic acid | |
(3-fluorophenyl)boronic acid |
Specifications
768-35-4 | |
100.0 | |
White | |
97% | |
FC6H4B(OH)2 | |
3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol, 3-fluorophenylbornic acid, fpba, boronic acid, b-3-fluorophenyl, 3-fluoro-phenyl-boronic acid | |
KNXQDJCZSVHEIW-UHFFFAOYSA-N | |
(3-fluorophenyl)boronic acid | |
2733986 | |
97% | |
3-Fluorophenylboronic acid |
96.0 | |
211.0°C to 212.0°C | |
Authentic | |
C6H6BFO2 | |
MFCD00236042 | |
Solubility in water: .. Other solubilities: soluble in methanol and thf | |
OB(O)C1=CC=CC(F)=C1 | |
139.92 | |
139.92 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/
GHS Signal Word: Warning
- 3-Fluorophenylboronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only