missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylacetic acid, 98%

Product Code. 11407537
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11407537 5 g 5g
11417537 25 g 25g
11427537 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11407537 Supplier Thermo Scientific Alfa Aesar Supplier No. A15635.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 331-25-9 | C8H7FO2 | 154.14 g/mol

3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.

Solubility
Slightly soluble in chloroform and methanol. Insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331-25-9
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004331
InChI Key YEAUYVGUXSZCFI-UHFFFAOYSA-N
Synonym 3-fluorophenylacetic acid, 2-3-fluorophenyl acetic acid, m-fluorophenylacetic acid, benzeneacetic acid, 3-fluoro, 3-fluorophenyl acetic acid, 3-fluorophenlacetic acid, acetic acid, m-fluorophenyl, m-fluorophenyl acetic acid, 3-fluorophenylaceticacid
PubChem CID 67617
IUPAC Name 2-(3-fluorophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(F)=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 151°C (0.8 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Quantity 5 g
Beilstein 775898
Solubility Information Slightly soluble in chloroform and methanol. Insoluble in water.
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 3-Fluorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.