missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylacetic acid, 98%, ACROS Organics™

Product Code. 10751721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10751721 5 g Glass bottle 5g
10481101 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10751721 Supplier Thermo Scientific Acros Supplier No. 278270050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 331-25-9
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004331
InChI Key YEAUYVGUXSZCFI-UHFFFAOYSA-N
Synonym 3-fluorophenylacetic acid, 2-3-fluorophenyl acetic acid, m-fluorophenylacetic acid, benzeneacetic acid, 3-fluoro, 3-fluorophenyl acetic acid, 3-fluorophenlacetic acid, acetic acid, m-fluorophenyl, m-fluorophenyl acetic acid, 3-fluorophenylaceticacid
PubChem CID 67617
IUPAC Name 2-(3-fluorophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(F)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 44°C to 48°C
Color White to Yellow or Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H4CH2COOH
Quantity 5 g
Beilstein 09, 2261
Formula Weight 154.14
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorophenylacetic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.