missing translation for 'onlineSavingsMsg'
Learn More

(3-Fluorophenyl)methanesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10306384
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10306384 5 g Amber Glass Bottle 5g
10718255 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10306384 Supplier Thermo Scientific Maybridge Supplier No. MO07830DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 24974-72-9
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.631
MDL Number MFCD04117456
InChI Key QKGIPEQAFOXGSI-UHFFFAOYSA-N
Synonym 3-fluorophenyl methanesulfonyl chloride, 3-fluoro-phenyl-methanesulfonyl chloride, 3-fluorophenyl methanesulphonyl chloride, benzenemethanesulfonyl chloride, 3-fluoro, chloro 3-fluorophenyl methyl sulfone, pubchem10052, 3-fluorobenzylsulphonyl chloride, benzenemethanesulfonylchloride,3-fluoro
PubChem CID 2759107
IUPAC Name (3-fluorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC(=C1)F)CS(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material (3-Fluorophenyl)methanesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.