missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenol, 98%

Product Code. 11460144
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460144 50 g 50g
11450144 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460144 Supplier Thermo Scientific Alfa Aesar Supplier No. A13598.18
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 372-20-3 | C6H5FO | 112.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-20-3
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.103
MDL Number MFCD00002254
InChI Key SJTBRFHBXDZMPS-UHFFFAOYSA-N
Synonym m-fluorophenol, phenol, 3-fluoro, 3-fluoro-phenol, phenol, m-fluoro, meta-fluorophenol, unii-t7oma38487, m-fluoro-phenol, 3-fluorphenol, 3-fluorophenol, 3-fluoro phenol
PubChem CID 9743
IUPAC Name 3-fluorophenol
SMILES C1=CC(=CC(=C1)F)O

Specifications

Melting Point 12°C to 14°C
Density 1.246
Boiling Point 177°C to 179°C
Flash Point 71°C (159°F)
Odor Phenol-like
Refractive Index 1.514
Quantity 50 g
UN Number UN2922
Beilstein 1904536
Formula Weight 112.1
Percent Purity 98%
Chemical Name or Material 3-Fluorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.