missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenethyl alcohol, 99%, Thermo Scientific Chemicals

Product Code. 10533231
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52059-53-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00045998
InChI Key MZNBGEKFZCWVES-UHFFFAOYSA-N
Synonym 2-3-fluorophenyl ethanol, 3-fluorophenethyl alcohol, 2-3-fluorophenyl ethan-1-ol, benzeneethanol, 3-fluoro, 3-fluorophenethylalcohol, 2-3-fluorophenyl ethylalcohol, 3-fluorobenzenethanol, 3-fluorophenylethanol, 3-fluorobenzeneethanol, acmc-209kxl
PubChem CID 573132
IUPAC Name 2-(3-fluorophenyl)ethanol
SMILES OCCC1=CC=CC(F)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.1200g/mL
Boiling Point 90°C (3.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2CH2OH
Refractive Index 1.5080 to 1.51
Quantity 5 g
Specific Gravity 1.12
Formula Weight 140.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Fluorophenethyl alcohol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'