missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenethyl alcohol, 99%, Thermo Scientific Chemicals

Product Code. 10270903 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10270903 1 g Glass bottle 1g
10533231 5 g Glass bottle 5g
10539643 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10270903 Supplier Thermo Scientific Acros Supplier No. 304690010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52059-53-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00045998
InChI Key MZNBGEKFZCWVES-UHFFFAOYSA-N
Synonym 2-3-fluorophenyl ethanol, 3-fluorophenethyl alcohol, 2-3-fluorophenyl ethan-1-ol, benzeneethanol, 3-fluoro, 3-fluorophenethylalcohol, 2-3-fluorophenyl ethylalcohol, 3-fluorobenzenethanol, 3-fluorophenylethanol, 3-fluorobenzeneethanol, acmc-209kxl
PubChem CID 573132
IUPAC Name 2-(3-fluorophenyl)ethanol
SMILES OCCC1=CC=CC(F)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.1200g/mL
Boiling Point 90°C (3.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2CH2OH
Refractive Index 1.5080 to 1.51
Quantity 1 g
Specific Gravity 1.12
Formula Weight 140.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Fluorophenethyl alcohol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.