missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl isocyanate, 98%

Product Code. 11945691 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11945691

Brand: Thermo Scientific Acros 437580010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 102422-56-0 | C8H6FNO | 151.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102422-56-0
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
InChI Key PHRJTGPFEAUEBC-UHFFFAOYSA-N
Synonym 3-fluorobenzyl isocyanate, 1-fluoro-3-isocyanatomethyl benzene, 3-fluorobenzylisocyanate, benzene,1-fluoro-3-isocyanatomethyl, benzene, 1-fluoro-3-isocyanatomethyl, pubchem16233, acmc-20aov0, 3-fluoro-benzyl isocyanate, 3-fluorophenylmethyl isocyanate
PubChem CID 643499
IUPAC Name 1-fluoro-3-(isocyanatomethyl)benzene
SMILES C1=CC(=CC(=C1)F)CN=C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Boiling Point 209.0°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 151.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzyl isocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Fluorobenzyl isocyanate, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.