missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl chloride, 97%

Product Code. 11460533
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11460533

Brand: Thermo Scientific Alfa Aesar A12665.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 456-42-8 | C7H6ClF | 144.573 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-42-8
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00000904
InChI Key XBDXMDVEZLOGMC-UHFFFAOYSA-N
Synonym 3-fluorobenzyl chloride, 1-chloromethyl-3-fluorobenzene, m-fluorobenzyl chloride, benzene, 1-chloromethyl-3-fluoro, 3-fluorobenzylchloride, alpha-chloro-3-fluorotoluene, alpha-chloro-m-fluorotoluene, fluorobenzyl-3 chloride, toluene, .alpha.-chloro-m-fluoro, .alpha.-chloro-m-fluorotoluene
PubChem CID 9974
IUPAC Name 1-(chloromethyl)-3-fluorobenzene
SMILES C1=CC(=CC(=C1)F)CCl

Specifications

Density 1.213
Boiling Point 176°C to 177°C
Flash Point 58°C (136°F)
Odor Pungent
Refractive Index 1.512
Quantity 5 g
UN Number UN2920
Beilstein 742265
Formula Weight 144.58
Percent Purity 97%
Chemical Name or Material 3-Fluorobenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.