missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl bromide, 95%

Product Code. 10220893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10220893 5 g Glass Bottle 5g
10091010 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10220893 Supplier Thermo Scientific Acros Supplier No. 119400050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 456-41-7 | C7H6BrF | 189.027 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-41-7
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00000340
InChI Key SCBZBMXPJYMXRC-UHFFFAOYSA-N
Synonym 3-fluorobenzyl bromide, 1-bromomethyl-3-fluorobenzene, 3-fluorobenzylbromide, m-fluorobenzyl bromide, benzene, 1-bromomethyl-3-fluoro, m-fluorobenzylbromide, alpha-bromo-m-fluorotoluene, alpha-bromo-3-fluorotoluene, toluene, .alpha.-bromo-m-fluoro, 1-bromobenzyl-3-fluorobenzene
PubChem CID 68007
IUPAC Name 1-(bromomethyl)-3-fluorobenzene
SMILES C1=CC(=CC(=C1)F)CBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.541g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Linear Formula FC6H4CH2Br
Refractive Index 1.5450 to 1.5480 (20°C, 589nm)
Quantity 5 g
Beilstein 05, II, 238
Formula Weight 189.03
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzyl bromide, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.