missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl alcohol, 98%, Thermo Scientific™

Product Code. 10563921 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10563921

Brand: Thermo Scientific Acros 171960250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 456-47-3
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00004631
InChI Key QDHRSLFSDGCJFX-UHFFFAOYSA-N
Synonym 3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486
PubChem CID 68008
IUPAC Name (3-fluorophenyl)methanol
SMILES C1=CC(=CC(=C1)F)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1600g/mL
Boiling Point 104°C to 105°C (22.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2OH
Refractive Index 1.5080 to 1.511
Quantity 25 g
Specific Gravity 1.16
Formula Weight 126.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzyl alcohol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.