missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl alcohol, 98%

Product Code. 11484154
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484154 10 g 10g
11339489 25 g 25g
11494154 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484154 Supplier Thermo Scientific Alfa Aesar Supplier No. A12509.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 456-47-3 | C7H7FO | 126.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-47-3
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00004631
InChI Key QDHRSLFSDGCJFX-UHFFFAOYSA-N
Synonym 3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486
PubChem CID 68008
IUPAC Name (3-fluorophenyl)methanol
SMILES C1=CC(=CC(=C1)F)CO

Specifications

Density 1.166
Boiling Point 201°C
Flash Point 90°C (194°F)
Odor Characteristic
Refractive Index 1.51
Quantity 10 g
Beilstein 2242511
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.