missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzoyl chloride, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -30°C |
| Density | 1.32 |
| Boiling Point | 188°C to 189°C |
| Flash Point | 82°C (179°F) |
| Odor | Pungent |
| Refractive Index | 1.5285 |
| Quantity | 25 g |
| UN Number | UN3265 |
| Beilstein | 636610 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
Product Identifier
- 3-Fluorobenzoyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: 3-Fluorobenzoyl chloride
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?