missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzonitrile, 98%

Product Code. 11495397
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11495397

Brand: Thermo Scientific Alfa Aesar B23426.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 403-54-3 | C7H4FN | 121.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-54-3
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001797
InChI Key JZTPKAROPNTQQV-UHFFFAOYSA-N
Synonym benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile
PubChem CID 67880
IUPAC Name 3-fluorobenzonitrile
SMILES FC1=CC=CC(=C1)C#N

Specifications

Melting Point -16°C
Density 1.14
Boiling Point 183°C
Flash Point 67°C (152°F)
Odor Almond-like
Refractive Index 1.5055
Quantity 100 g
UN Number UN3276
Beilstein 1858941
Formula Weight 121.11
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzonitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.