missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzonitrile, 98%, ACROS Organics™

Product Code. 10607022 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10607022

Brand: Thermo Scientific Acros 216580100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 403-54-3
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001797
InChI Key JZTPKAROPNTQQV-UHFFFAOYSA-N
Synonym benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile
PubChem CID 67880
IUPAC Name 3-fluorobenzonitrile
SMILES FC1=CC=CC(=C1)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1300g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H4CN
Refractive Index 1.5043 to 1.5063 (20°C, 589nm)
Quantity 10 g
Beilstein 09, II, 220
Formula Weight 121.11
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.