missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzoic hydrazide, 97%, Thermo Scientific™

Product Code. 10433170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10433170 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10433170 Supplier Thermo Scientific Acros Supplier No. 436340010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 499-55-8
CAS Min % 96.0
CAS Max % 100.0
Melting Point 140.0°C to 141.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C7H7FN2O
Quantity 1 g
Synonym 3-fluorobenzhydrazide, 3-fluorobenzoic hydrazide, 3-fluorobenzoic acid hydrazide, pubchem3482, acmc-20an3d, #, 3-fluorobenzenecarbohydrazide, ksc497s6p, 3-fluoro-benzoic acid hydrazide, 1-fluorobenzene-3-carbohydrazide
InChI Key UUISEXNUHLZEDB-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)F)C(=O)NN
IUPAC Name 3-fluorobenzohydrazide
Molecular Weight (g/mol) 154.15
PubChem CID 351640
Formula Weight 154.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorobenzoic hydrazide
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorobenzoic hydrazide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.