missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzoic Acid, TRC

Product Code. 30405556
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 30405556

Brand: Toronto Research Chemicals TRCF5883815G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Fluorobenzoic Acid
CAS 455-38-9
Molecular Formula C7H5FO2
Quantity 5 g
Synonym m-Fluorobenzoic Acid, NSC 10320, NSC 176125
InChI Formula InChI=1S/C7H5FO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,(H,9,10)
SMILES OC(=O)c1cccc(F)c1
IUPAC Name 3-fluorobenzoic acid
Molecular Weight (g/mol) 140.11
Formula Weight 140.0274
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.