missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzoic Acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3-Fluorobenzoic Acid |
| CAS | 455-38-9 |
| Molecular Formula | C7H5FO2 |
| Quantity | 1 g |
| Synonym | m-Fluorobenzoic Acid, NSC 10320, NSC 176125 |
| InChI Formula | InChI=1S/C7H5FO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,(H,9,10) |
| SMILES | OC(=O)c1cccc(F)c1 |
| IUPAC Name | 3-fluorobenzoic acid |
| Molecular Weight (g/mol) | 140.11 |
| Formula Weight | 140.0274 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction