missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzoic acid, 99%

Product Code. 11497827
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497827 25 g 25g
11407837 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497827 Supplier Thermo Scientific Alfa Aesar Supplier No. A16511.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 455-38-9 | C7H5FO2 | 140.11 g/mol

Used in the preparation of regulation of phenolic acid synthesis in Bacillus subtilis. Also used in the bacterial decarboxylation of o-phthalic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of regulation of phenolic acid synthesis in Bacillus subtilis. Also used in the bacterial decarboxylation of o-phthalic acids.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-38-9
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
MDL Number MFCD00002489
InChI Key MXNBDFWNYRNIBH-UHFFFAOYSA-N
Synonym m-fluorobenzoic acid, benzoic acid, 3-fluoro, meta-fluorobenzoic acid, benzoic acid, m-fluoro, 3-fluoro-benzoic acid, 3-fluorobenzoicacid, pubchem3480, 3-fluorobenzoic acid, ksc235q1b, paragos 390192
PubChem CID 9968
ChEBI CHEBI:20021
IUPAC Name 3-fluorobenzoic acid
SMILES OC(=O)C1=CC=CC(F)=C1

Specifications

Melting Point 122°C to 124°C
Density 1.474
Quantity 25 g
Beilstein 1906920
Solubility Information Very soluble in water.
Formula Weight 140.11
Percent Purity 99%
Chemical Name or Material 3-Fluorobenzoic acid
compliance-icons
Product Identifier
  • 3-Fluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Fluorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.