missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. 10078711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10078711 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10078711 Supplier Thermo Scientific Acros Supplier No. 356930010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

CAS 701-27-9
CAS Min % 97.5
CAS Max % 100.0
Boiling Point 108.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H4ClFO2S
Linear Formula FC6H4SO2Cl
Refractive Index 1.5280 to 1.5310
MDL Number MFCD00042286
Quantity 1g
Synonym 3-fluorobenzene-1-sulfonyl chloride, m-fluorobenzenesulfonyl chloride, 3-fluorobenzenesulfonylchloride, 3-fluorobenzenesulphonyl chloride, 3-fluoro-benzenesulfonyl chloride, 3-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 3-fluoro, pubchem5156, acmc-209od0, 3-fluorobenzensulfonylchloride
InChI Key OKYSUJVCDXZGKE-UHFFFAOYSA-N
SMILES FC1=CC(=CC=C1)S(Cl)(=O)=O
IUPAC Name 3-fluorobenzene-1-sulfonyl chloride
Molecular Weight (g/mol) 194.60
PubChem CID 2734258
Formula Weight 194.61
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzenesulfonyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.