missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15492068
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492068 5 g 5g
15402078 250 mg 250mg
15482068 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492068 Supplier Thermo Scientific Alfa Aesar Supplier No. H55740.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1524-40-9
Molecular Formula C6H6FNO2S
Molecular Weight (g/mol) 175.177
MDL Number MFCD00042284
InChI Key CRINBBOGNYCAOV-UHFFFAOYSA-N
Synonym 3-fluorobenzene-1-sulfonamide, benzenesulfonamide, 3-fluoro, 3-fluorobenzenesulphonamide, 3-fbsa, 2weo, pubchem11762, acmc-20a5kf, 3-fluoranylbenzenesulfonamide, ksc180k2p, 3-fluorobenzenesulfonamide
PubChem CID 150924
IUPAC Name 3-fluorobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)F

Specifications

Melting Point 124°C to 129°C
Color White to Brown
Quantity 5 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 175.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Fluorobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.