missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzamide, 99%, Thermo Scientific™

Product Code. 11945541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11945541 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11945541 Supplier Thermo Scientific Acros Supplier No. 184080100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 455-37-8
CAS Min % 98.5
CAS Max % 100.0
Melting Point 129°C to 132°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H6FNO
Linear Formula FC6H4CONH2
MDL Number MFCD00007983
Quantity 10 g
Beilstein 09, I, 137
Synonym m-fluorobenzamide, benzamide, 3-fluoro, 3-fluoro-benzamide, benzamide, m-fluoro, 3-fluoranylbenzamide, 3-fluorobenzamide, ksc492m7t, ypighniixyspkf-uhfffaoysa
InChI Key YPIGHNIIXYSPKF-UHFFFAOYSA-N
SMILES NC(=O)C1=CC=CC(F)=C1
IUPAC Name 3-fluorobenzamide
Molecular Weight (g/mol) 139.13
PubChem CID 68000
Formula Weight 139.13
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Fluorobenzamide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorobenzamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.