missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzamide, 99%

Product Code. 15468859
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 15468859

Brand: Thermo Scientific Alfa Aesar L06701.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 455-37-8 | C7H6FNO | 139.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 455-37-8
Melting Point 129°C to 133°C
Molecular Formula C7H6FNO
MDL Number MFCD00007983
Quantity 10 g
Beilstein 1859858
Synonym m-fluorobenzamide, benzamide, 3-fluoro, 3-fluoro-benzamide, benzamide, m-fluoro, 3-fluoranylbenzamide, 3-fluorobenzamide, ksc492m7t, ypighniixyspkf-uhfffaoysa
InChI Key YPIGHNIIXYSPKF-UHFFFAOYSA-N
SMILES NC(=O)C1=CC=CC(F)=C1
IUPAC Name 3-fluorobenzamide
Molecular Weight (g/mol) 139.13
PubChem CID 68000
Formula Weight 139.13
Percent Purity 99%
Chemical Name or Material 3-Fluorobenzamide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.